Cobalt(II) chloride hexahydrate 98-102%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

31277-100G
BJ31277-100GEA 345.78 USD
BJ31277-100G
Cobalt(II) chloride hexahydrate 98-102%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt: 86 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.005 %
Potassium (K) Max. 0.005 %
Copper (Cu) Max. 0.002 %
Magnesium (Mg) Max. 0.001 %
Manganese (Mn) Max. 0.005 %
Sodium (Na) Max. 0.01 %
Nickel (Ni) Max. 0.05 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.005 %
Nitrate (NO₃) Max. 0.01 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR