Potassium thiocyanate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Warning

31272-6X250G
BJ31272-250GCS 412.3 USD
BJ31272-250G BJ31272-6X250G
Potassium thiocyanate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Potassium thiocyanate
Formula: KSCN
Boiling Pt: Dec. (on heating)
Melting Pt: 172 °C
Density: 1.89 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now

Specification Test Results

Assay Min. 99 %
Water insoluble matter Max. 0.005 %
Insoluble in Ethanol Max. 0.01 %
pH (5 %, 25 °C) 5.3 - 8.5
Ammonium (NH₄) Max. 0.001 %
Copper (Cu) Max. 0.0002 %
Iron (Fe) Max. 1 ppm
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0002 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.001 %
Sulfide (S) Max. 0.001 %
Matter consuming I (as I) Max. 0.025 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR