Ammonium persulfate (APS) ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

31117-5KG
BJ31117-5KGEA 506.72 USD
BJ31117-5KG
Ammonium persulfate (APS) ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
Melting Pt: 120 °C
Density: 1.98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
UN: 1444
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Water insoluble matter Max. 0.005 %
Residue on ignition (SO₄) Max. 0.05 %
Titrable Acid Max. 0.04 meq/g
Iron (Fe) Max. 0.001 %
Manganese (Mn) Max. 0.00005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride, Chlorate (as Cl) Max. 0.001 %
Chloride (Cl) Max. 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR