Sodium bromide ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals

310506-500G
BJ310506-500GEA 138.37 USD
BJ310506-500G
Sodium bromide ≥99.0% ACS, Fluka™
Sodium bromide
Formula: NaBr
Boiling Pt: 1.393 °C (1.013 hPa)
Melting Pt: 750 °C
Density: 3.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (5 %, 25 °C) 5.0 - 8.8
Water insoluble matter Max. 0.005 %
Barium (Ba) Max. 0.002 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.1 %
Magnesium (Mg) Max. 0.001 %
Heavy metals (as Pb) Max. 5 ppm
Bromate (BrO₃) Max. 0.001 %
Chloride (Cl) Max. 0.2 %
Sulfate (SO₄) Max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR