tert-Butanol ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

308250-2.5L
BJ308250-1LEA 169.3 USD
BJ308250-1L BJ308250-2.5L
tert-Butanol ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
tert-Butanol

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: (CH₃)₃COH
Boiling Pt: 81 - 83 °C
Melting Pt: 25 °C
Density: 0.78 g/cm3
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.50 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Absorbance at 350 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 250 nm Max. 0.20
Absorbance at 230 nm Max. 0.50
Absorbance at 215 nm Max. 1.00
Explosion Limit 8 %
Refractive Index n20/D 1.387 (lit.)
Vapor Pressure 44 mmHg ( 26 °C)
Autoignition Temperature 896 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR