1,2-Dimethoxyethane ≥99.9% inhibitor-free, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

307432-1L
BJ307432-1LEA 340.87 USD
BJ307432-1L
1,2-Dimethoxyethane ≥99.9% inhibitor-free, CHROMASOLV™ for HPLC, Riedel-de Haën™
1,2-Dimethoxyethane

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: C₄H₁₀O₂
Boiling Pt: 85 °C (1.013 hPa)
Melting Pt: -58 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: -2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008502
CAS Number: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3,II

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.85 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.03 %
Peroxides (as H₂O₂) Max. 15 ppm
Absorbance at 400 nm Max. 0.01
Absorbance at 350 nm Max. 0.01
Absorbance at 300 nm Max. 0.03
Absorbance at 250 nm Max. 0.20
Absorbance at 220 nm Max. 1.0
Vapor Density 3.1 (20 °C, vs air)
Refractive Index n20/D 1.379 (lit.)
Explosion Limit 10.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR