Dichloromethane ≥99.9% stabilized, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Methane dichloride, Methylene chloride, Methylene chloride

Stabilization: Stabilised with amylene

299-4 CS299AA-200
BJ299-4CS 1070 USD
BJ299-4 BJCS299AA-200
Dichloromethane ≥99.9% stabilized, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Dichloromethane

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 40 °C
Melting Pt: –95 °C
Density: 1.33 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity (as HCl) 1 mg/L max.
Chloride 10 mg/L max.
Electron Capture GC 10 ng/L max.
Flame Ionization GC (as nonane) 5 ppb max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.4236 - 1.4246
Residue 1 mg/L max.
UV Cutoff 233 nm max.
Water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR