4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

293261-1L
BJ293261-1LEA 142.04 USD
BJ293261-1L
4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
4-Methyl-2-pentanone

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 115…117 °C (1.013 hPa)
Melting Pt: –83 °C
Density: 0.8 g/cm³ (20 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.50 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Absorbance at 400 nm Max. 0.01
Absorbance at 380 nm Max. 0.02
Absorbance at 360 nm Max. 0.15
Absorbance at 340 nm Max. 0.50
Absorbance at 335 nm Max. 1.0
Explosion Limit 1.2-8 %, 93 °F
Vapor Density 3.5 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR