Molecular sieve dehydrate, beads for drying gases, with indicator, Fluka™
Supplier: Honeywell Research Chemicals
28211-1KG
BJ28211-1KGEA 910 USD
BJ28211-1KG BJ28211-250G
Molecular sieve dehydrate, beads for drying gases, with indicator, Fluka™
Molecular sieve dehydrate
Order Now
Specification Test Results
Appearance (color) complying
Loss on ignition (900°C) Max. 4.0 %
Water adsorption capacity Min. 15 %