Alcohol, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

270741-4X4L
BJ270741-4LCS 1320 USD
BJ270741-4L
Alcohol, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Alcohol
Formula: H₃CCH₂OH
Boiling Pt: 78.32 °C (main component)
Melting Pt: –114.1 °C (main component)
Density: 0.7890 g/cm³ (20 °C)
Flash Pt: 15 °C (main component)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 89.0 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.1 %
Methanol (GC) 3.0 - 5.0 %
2-propanol (GC) 4.0 - 6.0 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 255 nm Max. 0.04
Absorbance at 220 nm Max. 0.35
Absorbance at 210 nm Max. 0.65
Absorbance at 205 nm Max. 1.00


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR