Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Synonyms:
Dihydrogen oxide
270733-2.5L
270733-4L
270733-6X1L
BJ270733-1LCS
539.48
USD
BJ270733-1L
BJ270733-2.5L
BJ270733-4L
BJ270733-6X1L
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Water
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Formula:
H₂O Boiling Pt: 100 °C Melting Pt: 0 °C Density: 0.9971 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 |
Specification Test Results
Non-volatile matter | Max. 0.0003 % |
Absorbance at 400 nm | Max. 0.005 |
Absorbance at 250 nm | Max. 0.005 |
Absorbance at 210 nm | Max. 0.01 |
Absorbance at 205 nm | Max. 0.01 |
HPLC Gradient at 254 nm | Max. 2 mAU |
HPLC Gradient at 210 nm | Max. 10 mAU |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...