Pyridine ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Azabenzene, Azine

270407-1L
BJ270407-1LEA 413.35 USD
BJ270407-1L
Pyridine ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Pyridine
Formula: C₅H₅N
Boiling Pt: 115 °C (1.013 hPa)
Melting Pt: -42 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 12.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

IR spectrum complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0004 %
Water (Karl Fischer) Max. 0.05 %
Absorbance at 400 nm Max. 0.01
Absorbance at 350 nm Max. 0.01
Absorbance at 335 nm Max. 0.02
Absorbance at 315 nm Max. 0.15
Absorbance at 305 nm Max. 1.0
Autoignition Temperature 899 °F
Explosion Limit 12.4 %
Refractive Index n20/D 1.509 (lit.)
Vapor Pressure 10 mmHg ( 13.2 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR