Tetrachloroethylene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene

270393-1L
BJ270393-1LEA 257.52 USD
BJ270393-1L
Tetrachloroethylene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Tetrachloroethylene

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: C₂Cl₄
MW: 165.83 g/mol
Boiling Pt: 120-122 °C (1.013 hPa)
Melting Pt: -22 °C
Density: 1.622…1.624 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Absorbance at 400 nm Max. 0.03
Absorbance at 350 nm Max. 0.05
Absorbance at 305 nm Max. 0.10
Absorbance at 300 nm Max. 0.20
Absorbance at 295 nm Max. 0.30
Absorbance at 290 nm Max. 1.0
APHA Max. 10
Vapor Pressure 13 mmHg ( 20 °C)
Vapor Density 5.83 (vs air)
Refractive Index n20/D 1.505 (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR