Formic acid 98.0-100%, Puriss., meets analytical specifications of DAC, FCC, Fluka™

Supplier: Honeywell Research Chemicals
Danger

27001-70KG
BJ27001-70KGEA 827.12 USD
BJ27001-70KG
Formic acid 98.0-100%, Puriss., meets analytical specifications of DAC, FCC, Fluka™
Formic acid
Formula: HCO₂H
Boiling Pt: 100.4 °C (1.013 hPa)
Melting Pt: 4 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 49.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
Identity complying
Assay 98.0 - 100.5 %
Density (D 20/20) 1.217 - 1.223
Non-volatile matter Max. 0.005 %
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 10 ppm
Sulfate (SO₄) Max. 40 ppm
Sulfite (SO₃) complying
Acetic acid (GC) Max. 0.05 %
Solubility in water complying
Residual solvents complying
Chromatographic purity complying
Refractive Index n20/D 1.370 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR