Soda lime, granular ACS +100 mesh, Fluka™

Supplier: Honeywell Research Chemicals
Danger

266434-500G
BJ266434-500GEA 155.16 USD
BJ266434-500G
Soda lime, granular ACS +100 mesh, Fluka™
Soda lime
Storage Temperature: Ambient
MDL Number: MFCD00134094
CAS Number: 8006-28-8
UN: 1907
ADR: 8,III

Order Now

Specification Test Results

Form granular
Loss on drying (200°C) Max. 7 %
Grain size < 100 mesh Max. 1 %
CO₂-absorption capacity Min. 19.0 %
Capacity ≥19.0 % absorption capacity (CO₂)
Particle Size +100 mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR