Methylcyclohexane 99%, Spectrophotometric Grade, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

259691-1L 259691-2L
BJ259691-1LEA 339 USD
BJ259691-1L BJ259691-2L
Methylcyclohexane 99%, Spectrophotometric Grade, Riedel-de Haën™
Methylcyclohexane
Formula: C₆H₁₁CH₃
Boiling Pt: 100-102 °C (1.013 hPa)
Melting Pt: -126 °C
Density: 0.769 g/cm³ (20 °C)
Flash Pt: 6.7 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 98.5 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 260 nm Max. 0.05
Absorbance at 232 nm Max. 0.15
Absorbance at 221 nm Max. 0.40
Absorbance at 207 nm Max. 1.0
Explosion Limit 6.7 %
Vapor Pressure 83.2 mmHg ( 37.7 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR