Sodium formate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Formic acid sodium salt

247596-500G
BJ247596-500GEA 136.51 USD
BJ247596-500G
Sodium formate ≥99.0% ACS, Fluka™
Sodium formate
Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 255 °C
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0

Order Now

Specification Test Results

Identity (IR) complying
Assay Min. 99.0 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR