Charcoal activated, powder, DARCO® -100 mesh, Fluka™

Supplier: Honeywell Research Chemicals
Warning

242276-1KG
BJ242276-1KGEA 249.74 USD
BJ242276-1KG
Charcoal activated, powder, DARCO® -100 mesh, Fluka™
Charcoal activated

  • DARCO is a registered trademark of Norit Americas, Inc.

Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3

Order Now

Specification Test Results

Particle Size −100 mesh
Appearance complying
Loss on drying (105°C) Max. 12 %
Methylene blue adsorption Min. 15 g/100g
Water soluble matter Max. 0.50 %
In acid soluble iron (Fe) Max. 200 ppm
Vapor Pressure <0.1 mmHg ( 20 °C)
Resistivity 1375 μΩ-cm, 20°C (graphite)
Form powder
Autoignition Temperature 842 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR