(±)-2-Butanol ≥99%

Supplier: Honeywell Research Chemicals
Warning

24126-30KG
BJ24126-30KGEA 783.96 USD
BJ24126-30KG
(±)-2-Butanol ≥99%
(±)-2-Butanol
Formula: H₃CCH₂CH(OH)CH₃
Boiling Pt: 99 °C
Melting Pt: –89 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
UN: 1120
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=010e04e8f508fd3d501873d20c2349817834b6e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR