Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning

24122-30KG 24122-1L 24122-2.5L 24122-4X2.5L 24122-6X1L
BJ24122-30KGEA 1178.74 USD
BJ24122-30KG BJ24122-1L BJ24122-2.5L BJ24122-4X2.5L BJ24122-6X1L
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Benzyl alcohol

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 878307
RTECS Number: DN3150000
UNSPSC Code: 12191503

Formula: C₆H₅CH₂OH
Boiling Pt: 205.3 °C (1.013 hPa)
Melting Pt: –15.4 °C
Density: 1.045 g/cm³ (20 °C)
Flash Pt: 100.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ignition Residue ≤0.005% (as SO₄)
Identity (IR) complying
Assay (GC) 99.0 - 100.5 %
Density (D 20/20) 1.043 - 1.049
Density (D 25/25) 1.042 - 1.047
Refractive index (n 20/D) 1.539 - 1.541
Non-volatile matter Max. 0.05 %
Water (Karl Fischer) Max. 0.1 %
Sulfated ash Max. 0.005 %
Free acid complying
Free acid (as C₆H₅COOH) Max. 0.02 %
Benzaldehyde (GC) Max. 0.05 %
Cyclohexylmethanol (GC) Max. 0.10 %
Sum peaks before Benzyl alcohol (GC) Max. 0.02 %
Sum peaks after Benzyl alcohol (GC) Max. 0.2 %
Related substances (GC) complying
Peroxide value Max. 5
Appearance of the solution complying
Residual solvents complying
Vapor Density 3.7 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR