Sodium sulfite ≥98.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals

239321-500G
BJ239321-500GEA 66.31 USD
BJ239321-500G
Sodium sulfite ≥98.0% ACS, Fluka™
Sodium sulphite
Formula: Na₂SO₃
Melting Pt: 600 °C
Density: 2.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4

Order Now

Specification Test Results

Assay (Iodometric) Min. 98.0 %
Water insoluble matter Max. 0.005 %
Free acid complying
Titrable Base Max. 0.03 meq/g
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR