Sodium hypochlorite 4.00 - 4.99% available chlorine in aqueous solution, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

239305-500ML
BJ239305-500MLEA 89.28 USD
BJ239305-500ML BJ239305-5L
Sodium hypochlorite 4.00 - 4.99% available chlorine in aqueous solution, Reagent Grade
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Boiling Pt: 111 °C
Melting Pt: –30…–20 °C
Density: 1.1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,III

Order Now

Specification Test Results

Assay of active chlorine 4 - 5 %
Vapor Pressure 17.5 mmHg ( 20 °C)
Composition available chlorine, 4.00-4.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR