Sodium hexanitritocobaltate(III) ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

233722-100G 233722-500G
BJ233722-100GEA 212.56 USD
BJ233722-100G BJ233722-500G
Sodium hexanitritocobaltate(III) ACS, Fluka™
Sodium hexanitritocobaltate(III)
Formula: CoN₆Na₃O₁₂
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00003512
CAS Number: 13600-98-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance of the substance complying
Insoluble in acetic acid Max. 0.020 %
Suitability for K determination complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR