Sodium carbonate monohydrate ≥99.5% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

230952-500G
BJ230952-100GEA 44.78 USD
BJ230952-100G BJ230952-2.5KG BJ230952-500G
Sodium carbonate monohydrate ≥99.5% ACS, Fluka™
Sodium carbonate monohydrate
Formula: Na₂CO₃·1H₂O
Melting Pt: 851 °C
Density: 2.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149177
CAS Number: 5968-11-6

Order Now

Specification Test Results

Assay Min. 99.5 %
Loss on drying (150°C) 13.0 - 15.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.03 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 5 ppm
S-compounds (as SO₄) Max. 0.004 %
Silicate (as SiO₂) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR