Ammonium dihydrogen phosphate ≥98% ACS, Fluka™

Supplier: Honeywell Research Chemicals

216003-500G
BJ216003-100GEA 50.6 USD
BJ216003-100G BJ216003-2.5KG BJ216003-500G
Ammonium dihydrogen phosphate ≥98% ACS, Fluka™
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
Boiling Pt: 197 °C (dec.)
Melting Pt: 190 °C
Density: 1.803 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5

Order Now

Specification Test Results

Identity (IR) complying
Assay Min. 98.0 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 3.8 - 4.4
Calcium (Ca) Max. 0.001 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 5 ppm
Sodium (Na) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Nitrate (NO₃) Max. 0.001 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR