Mercury (II) oxide, red ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

213357-100G
BJ213357-100GEA 162.2 USD
BJ213357-100G
Mercury (II) oxide, red ≥99.0% ACS, Fluka™
Mercury (II) oxide
Formula: HgO
Melting Pt: 400 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011045
CAS Number: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1,II

Order Now

Specification Test Results

Assay Min. 99.0 %
Insoluble in HCl Max. 0.03 %
Residue on ignition a. reducing with HCOOH (SO₄)* Max. 0.025 %
Iron (Fe) Max. 0.005 %
Chloride (Cl) Max. 0.025 %
Sulfate (SO₄) Max. 0.015 %
Total N Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR