Hexane (mixture of isomers) ≥95% (n-hexane) ≥99.9% (by GC) (n-Hexane > 95%), B&J Brand™ for gas chromatography, for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hexanes, Isohexane

213-4 CS213-200 CS213-56
BJ213-4CS 2271 USD
BJ213-4 BJCS213-200 BJCS213-56
Hexane (mixture of isomers) ≥95% (n-hexane) ≥99.9% (by GC) (n-Hexane > 95%), B&J Brand™ for gas chromatography, for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™
Hexane (mixture of isomers)

Suitable for gas chromatography and pesticide residue analysis.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68.7 °C
Melting Pt: –95 °C
Density: 0.659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10 max.
Electron Capture GC (as lindane) 10 ng/L max.
Flame Ionization GC (as nonane) 5 ppb max.
GC Analysis (n-Hexane) 95 % min.
GC Analysis (total C6 isomers) 99.9 % min.
Residue 1 mg/L max.
Thiophene Passes test
Water by Karl Fisher Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR