Calcium hydroxide ≥96.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Slaked lime

21181-1KG 21181-5KG
BJ21181-1KGEA 74.57 USD
BJ21181-1KG BJ21181-5KG
Calcium hydroxide ≥96.0% (by titrimetric analysis), Purum p.a.
Calcium hydroxide

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: Ca(OH)₂
Boiling Pt: Decomposes on heating
Melting Pt: 580 °C
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3

Order Now

Specification Test Results

Assay 96.0 - 102.0 %
Carbonate (as CaCO₃) Max. 4 %
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 500 ppm
Potassium (K) Max. 200 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 500 ppm
Chloride (Cl) Max. 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR