Molecular sieve A4 (0.4 nm, 4 Å), beads 4 - 8 mesh, Fluka™
Supplier: Honeywell Research Chemicals
208590-1KG
208590-5KG
BJ208590-1KGEA
234
USD
BJ208590-1KG
BJ208590-500G
BJ208590-5KG
Molecular sieve A4 (0.4 nm, 4 Å), beads 4 - 8 mesh, Fluka™
Molecular sieve A4 (0.4 nm, 4 Å)
Melting Pt:
400 °C Density: 2.0 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD03457537 CAS Number: 70955-01-0 |
Specification Test Results
Form | beads |
Grain size 4 - 8 mesh | Min. 95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...