Molecular sieve A4 (0.4 nm, 4 Å), beads 4 - 8 mesh, Fluka™

Supplier: Honeywell Research Chemicals

208590-1KG 208590-5KG
BJ208590-1KGEA 234 USD
BJ208590-1KG BJ208590-500G BJ208590-5KG
Molecular sieve A4 (0.4 nm, 4 Å), beads 4 - 8 mesh, Fluka™
Molecular sieve A4 (0.4 nm, 4 Å)
Melting Pt: 400 °C
Density: 2.0 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR