Potassium thiocyanate ≥99.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Potassium rhodanide
207799-100G
207799-2.5KG
BJ207799-100GEA
110
USD
BJ207799-100G
BJ207799-2.5KG
Potassium thiocyanate ≥99.0% ACS, Fluka™
Potassium thiocyanate
Formula:
KSCN Boiling Pt: Decomposes on heating Melting Pt: 172 °C Density: 1.89 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 |
Specification Test Results
Assay | Min. 98.5 % |
Water insoluble matter | Max. 0.005 % |
pH (5 %, 25 °C) | 5.3 - 8.7 |
Ammonium (NH₄) | Max. 3 ppm |
Iron (Fe) | Max. 2 ppm |
Sodium (Na) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.005 % |
Sulfate (SO₄) | Max. 0.005 % |
Matter consuming I (as I) | complying |
IR spectrum | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...