tert-Butanol ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

19460-1L 19460-2.5L 19460-500ML 19460-5L
BJ19460-1LEA 198 USD
BJ19460-1L BJ19460-2.5L BJ19460-500ML BJ19460-5L
tert-Butanol ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
tert-Butanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: (CH₃)₃COH
Melting Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Identity (NMR) complying
Assay (GC) Min. 99.7 %
Solidification Point 24.0 - 25.5 °C
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.1 %
Titrable Acid Max. 0.001 meq/g
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Carbonyl Compounds (as HCHO) Max. 100 ppm
APHA Max. 20
Vapor Pressure 31 mmHg ( 20 °C)
Refractive Index n20/D 1.387 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR