Acetonitrile : water : trifluoroacetic acid (50 : 47.5 : 2.5) %, LabReady™, Solvent Blend for mass spectrometry, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

19182-250ML
BJ19182-250MLEA 168 USD
BJ19182-250ML
Acetonitrile : water : trifluoroacetic acid (50 : 47.5 : 2.5) %, LabReady™, Solvent Blend for mass spectrometry, Riedel-de Haën™
Acetonitrile : water : trifluoroacetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Flash Pt: <10 °C
Storage Temperature: Ambient
CAS Number: 75-05-8, 7732-18-5, 76-05-1
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Assay trifluoroacetic acid 2.0 - 3.0 %
Water (Karl Fischer) 47 - 49 %
Barium (Ba) Max. 5 ppm
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 20 ppm
Potassium (K) Max. 20 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 20 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR