Hydrobromic acid ≥48%, Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

18730-1L
BJ18730-1LEA 156.3 USD
BJ18730-1L BJ18730-250ML
Hydrobromic acid ≥48%, Purum p.a.
Hydrogen bromide

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: HBr
Boiling Pt: 126 °C (1.013 hPa)
Melting Pt: –11 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now

Specification Test Results

Assay 48.0 - 50.0 %
Refractive index (n 20/D) 1.440 - 1.443
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Sulfate and sulfite (as SO₄) Max. 50 ppm
Vapor Pressure 320 psi ( 21.1 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR