Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

184519-2.5L 184519-1L 184519-6X1L
BJ184519-2.5LEA 161.04 USD
BJ184519-2.5L BJ184519-1L BJ184519-6X1L
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Petroleum spirit

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


UNSPSC Code: 12191502

Boiling Pt: 40…60 °C (1.013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Identity (IR) complying
Refractive index (n 20/D) 1.358 - 1.368
Non-volatile matter Max. 0.001 %
Boiling range 35 - 60 °C
Explosion Limit 8 %
Acidity complying
APHA Max. 10
Autoignition Temperature 475 °F
Vapor Pressure 7.99 psi ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR