Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
(Diethylamino)ethane, TEA
17924-10X2ML
17924-1L
BJ17924-10X2MLCS
189
USD
BJ17924-10X2ML
BJ17924-1L
Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™
Triethylamine
Beilstein Number: 605283
RTECS Number: YE0175000
UNSPSC Code: 12352116
Formula:
(C₂H₅)₃N Boiling Pt: 90 °C Melting Pt: –115 °C Density: 0.73 g/cm³ (20 °C) Flash Pt: –11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009051 CAS Number: 121-44-8 EINECS: 204-469-4 UN: 1296 ADR: 3,II |
|
Specification Test Results
Assay (GC) | min. 99.5 % |
Suitability test (HPLC) | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...