Ammonium formate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Formic acid ammonium salt
17843-250G
17843-50G
BJ17843-250GEA
755
USD
BJ17843-250G
BJ17843-50G
Ammonium formate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Ammonium formate
Formula:
HCO₂NH₄ MW: 63.06 g/mol Melting Pt: 116 °C Density: 1.266 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013103 CAS Number: 540-69-2 EINECS: 208-753-9 Merck Index: 13,00525 |
Specification Test Results
Appearance of the solution (c=10%) | complying |
Assay (calculated on the anhydrous basis) | 99.0 - 101.0 % |
Water (Karl Fischer) | Max. 2 % |
pH (10 %, 20°C) | 5.5 - 7.5 |
Absorbance at 250 nm | Max. 0.05 |
Absorbance at 260 nm | Max. 0.04 |
Absorbance at 270 nm | Max. 0.03 |
Absorbance 280 nm | Max. 0.02 |
Absorbance 500 nm | Max. 0.02 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...