Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™

Supplier: Honeywell Research Chemicals

17836-250G 17836-50G
BJ17836-250GEA 376.16 USD
BJ17836-250G BJ17836-50G
Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Ammonium acetate
Formula: H₃CCOONH₄
Melting Pt: 113 °C
Density: 1.170 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Solid
assay 68.0 - 69.0 %
silver (Ag) Max. 10 ppb
aluminium (Al) Max. 20 ppb
gold (Au) Max. 10 ppb
arsenic (As) Max. 5 ppb
boron (B) Max. 10 ppb
barium (Ba) Max. 10 ppb
beryllium (Be) Max. 10 ppb
bismuth (Bi) Max. 10 ppb
calcium (Ca) Max. 30 ppb
cadmium (Cd) Max. 10 ppb
cobalt (Co) Max. 10 ppb
chromium (Cr) Max. 30 ppb
copper (Cu) Max. 10 ppb
iron (Fe) Max. 100 ppb
gallium (Ga) Max. 10 ppb
germanium (Ge) Max. 10 ppb
indium (In) Max. 10 ppb
potassium (K) Max. 50 ppb
lithium (Li) Max. 10 ppb
magnesium (Mg) Max. 50 ppb
manganese (Mn) Max. 10 ppb
molybdenum (Mo) Max. 10 ppb
sodium (Na) Max. 50 ppb
nickel (Ni) Max. 30 ppb
lead (Pb) Max. 10 ppb
platinum (Pt) Max. 10 ppb
antimony (Sb) Max. 5 ppb
tin (Sn) Max. 10 ppb
strontium (Sr) Max. 10 ppb
titanium (Ti) Max. 50 ppb
thallium (Tl) Max. 10 ppb
vanadium (V) Max. 10 ppb
zinc (Zn) Max. 10 ppb
zirconium (Zr) Max. 10 ppb
chloride (Cl) Max. 0.5 ppm
phosphate (PO4) Max. 0.5 ppm
sulfate (SO4) Max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR