Tetrachloroethylene ≥99.5% stabilized

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene

16211-2.5L
BJ16211-1LEA 122.13 USD
BJ16211-1L BJ16211-2.5L BJ16211-6X1L
Tetrachloroethylene ≥99.5% stabilized
Tetrachloroethylene

We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:


  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.


Beilstein Number: 1361721
RTECS Number: KX3850000
UNSPSC Code: 12190000

Formula: C₂Cl₄
MW: 165.83 g/mol
Boiling Pt: 120…122 °C (1.013 hPa)
Melting Pt: –22 °C
Density: 1.622…1.624 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 5.83 (vs air)
Contains stabilizer
Assay Min. 99.5 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.001 %
Refractive Index n20/D 1.505 (lit.)
Vapor Pressure 19 mmHg ( 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR