(±)-2-Methyltetrahydrofuran ≥99.5% stabilized, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 150-400 ppm

155810-2.5L 155810-4L
BJ155810-100MLEA 106.28 USD
BJ155810-100ML BJ155810-2.5L BJ155810-500ML BJ155810-4L
(±)-2-Methyltetrahydrofuran ≥99.5% stabilized, Reagent Grade
(±)-2-Methyltetrahydrofuran

  • ≥95% for general laboratory use.


Beilstein Number: 102448
RTECS Number: LU2800000
UNSPSC Code: 12190000

Formula: C₅H₁₀O
Boiling Pt: 78-80 °C (1.013 hPa)
Melting Pt: <-20 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: -10 °C
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3,II

Order Now

Specification Test Results

Assay (GC) Min. 99.50 %
Assay BHT (GC) 150 - 400 ppm
Water (Karl Fischer) Max. 1.0 %
IR spectrum complying
Peroxides (as H₂O₂) Max. 100 ppm
Contains 150-400 ppm BHT as stabilizer
Refractive Index n20/D 1.406 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR