Lead(II) oxide, yellow ≥99.0% (by complexometric analysis), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

15338-1KG
BJ15338-1KGEA 231.4 USD
BJ15338-1KG BJ15338-250G BJ15338-50G
Lead(II) oxide, yellow ≥99.0% (by complexometric analysis), Puriss. p.a., Fluka™
Lead (II) oxide
Formula: PbO
Boiling Pt: 1.470 °C (1.013 hPa)
Melting Pt: 890 °C
Density: 9.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Assay (complexometric) 99.0 - 101.0 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 20 ppm
Total N Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR