Zinc oxide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals

14439-6X1KG
BJ14439-100GEA 53.14 USD
BJ14439-100G BJ14439-1KG
Zinc oxide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Zinc oxide
Formula: ZnO
Melting Pt: 1.975 °C
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5

Order Now

Specification Test Results

Assay (calculated to ignited substance) 99.0 - 100.5 %
Loss on ignition (500°C) Max. 1.0 %
Alkalinely reacting substances complying
Arsenic (As) Max. 3 ppm
Cadmium (Cd) Max. 10 ppm
Iron (Fe) Max. 10 ppm
Lead (Pb) Max. 50 ppm
Lead complying
Iron and other heavy metals complying
Carbonate and color of the solution complying
Carbonates and substances insoluble in acids complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR