Zinc chloride 98-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals
Danger

14422-25KG
BJ14422-1KGCS 1251.06 USD
BJ14422-1KG BJ14422-25KG BJ14422-500G
Zinc chloride 98-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Formula: ZnCl₂
Melting Pt: 287…304 °C (1.013 hPa)
Density: 2.93 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR