Sodium tungstate dihydrate ≥99%, Purum, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate

14304-6X500G
BJ14304-500GCS 2399.51 USD
BJ14304-500G
Sodium tungstate dihydrate ≥99%, Purum, Fluka™
Sodium wolframate dihydrate
Formula: Na₂WO₄·2H₂O
Melting Pt: 698 °C
Density: 4.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2

Order Now

Specification Test Results

Assay Min. 99 %
Free alkali (as NaOH) Max. 0.1 %
Iron (Fe) Max. 0.002 %
Molybdenum (Mo) Max. 0.01 %
Chloride (Cl) Max. 0.01 %
Sulfate (SO₄) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR