Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

14282-2L
BJ14282-2LEA 257 USD
BJ14282-2L
Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Water with 0,1% (v/v) acetic acid

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise

Density: 1.000 g/cm³
Storage Temperature: Ambient
EINECS: 231-791-2

Order Now

Specification Test Results

Assay of acetic acid 0.093 - 0.107 %
pH (20 °C) 3.2 - 3.5
Calcium (Ca) Max. 0.2 ppm
Potassium (K) Max. 0.2 ppm
Magnesium (Mg) Max. 0.2 ppm
Sodium (Na) Max. 0.5 ppm
Transmittance at 254 nm Min. 99 %
Transmittance at 230 nm Min. 75 %
Transmittance at 210 nm Min. 20 %
UHPLC-MS ESI(+) (Reserpine) Max. 10 ppb
UHPLC-MS ESI(-) (Digoxin) Max. 50 ppb
Suitability for the UHPLC-MS complying
LC-MS application complying
HPLC Gradient at 254 nm Max. 2 mAU
HPLC Gradient at 210 nm Max. 20 mAU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR