Water with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

14281-2L
BJ14281-2LEA 253 USD
BJ14281-2L
Water with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Water with 0,1% (v/v) formic acid
Density: 0.998 g/cm³ (20 °C)
Storage Temperature: Ambient
EINECS: 231-791-2

Order Now

Specification Test Results

Assay of formic acid 0.093 - 0.107 %
pH (20 °C) 2.6 - 2.8
Calcium (Ca) Max. 0.2 ppm
Potassium (K) Max. 0.2 ppm
Magnesium (Mg) Max. 0.2 ppm
Sodium (Na) Max. 0.5 ppm
Transmittance at 210 nm Min. 5 %
Transmittance at 230 nm Min. 45 %
Transmittance at 254 nm Min. 98.5 %
UHPLC-MS ESI(+) (Reserpine) Max. 10 ppb
UHPLC-MS ESI(-) (Digoxin) Max. 50 ppb
HPLC Gradient at 210 nm Max. 20 mAU
HPLC Gradient at 254 nm Max. 5 mAU
Suitability for the UHPLC-MS complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR