Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

14272-2L
BJ14272-2LEA 621 USD
BJ14272-2L
Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Acetonitrile with 0,1% (v/v) Formic acid
Boiling Pt: 81 °C
Melting Pt: –46 °C
Density: 0.78 g/cm³ (20 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
EINECS: 200-835-2
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Formic acid 0.095 - 0.105 Vol.%
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.02 %
Aluminium (Al) Max. 0.03 ppm
Calcium (Ca) Max. 0.1 ppm
Iron (Fe) Max. 0.05 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.03 ppm
Sodium (Na) Max. 0.1 ppm
Transmittance at 254 nm Min. 90 %
Transmittance at 230 nm Min. 15 %
Transmittance at 210 nm Min. 5 %
Suitability for the UHPLC-MS complying
UHPLC Gradient drift 254 nm Max. 30 mAU
UHPLC Gradient peak 254 nm Max. 2 mAU
UHPLC-MS ESI(+) (Reserpine) Max. 20 ppb
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
Fluorescence (chinin) at 254 nm Max. 0.5 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR