1,2,4-Trichlorobenzene ≥99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning

132047-2.5L
BJ132047-2.5LEA 172.46 USD
BJ132047-2.5L
1,2,4-Trichlorobenzene ≥99%, Reagent Grade
1,2,4-Trichlorobenzene
Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 213.5 °C
Melting Pt: 17 °C
Density: 1.454 g/cm³ (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1,III

Order Now

Specification Test Results

Vapor Pressure 1 mmHg ( 40 °C)
Identity (IR) complying
Assay (GC) Min. 99 %
Refractive Index n20/D 1.571 (lit.)
Vapor Density >6 (vs air)
Explosion Limit 6.6 %, 150 °F

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR