Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™

Supplier: Honeywell Research Chemicals
Danger

13020-500G
BJ13020-100GEA 66.43 USD
BJ13020-100G BJ13020-100KG BJ13020-1KG BJ13020-500G
Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™
Lithium hydroxide monohydrate
Formula: LiOH·1H₂O
Melting Pt: 423 °C
Density: 1.830 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II

Order Now

Specification Test Results

Assay Min. 98.5 %
Assay of Li₂CO₃ Max. 1 %
Calcium (Ca) Max. 0.05 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.02 %
Sodium (Na) Max. 0.02 %
Chloride (Cl) Max. 0.05 %
Sulfate (SO₄) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR