1-Octanol 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning

112615-2.5L
BJ112615-2.5LEA 189.64 USD
BJ112615-2.5L BJ112615-20L
1-Octanol 99%, Reagent Grade
Formula: H₃C(CH₂)₆CH₂OH
Boiling Pt: 194 °C (1.013 hPa)
Melting Pt: –15 °C
Density: 0.827 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 98.5 %
Refractive index (n 20/D) 1.428 - 1.431
Non-volatile matter Max. 0.1 %
Water (Karl Fischer) Max. 0.5 %
Free acid (as CH₃COOH) Max. 0.01 %
Chloride (Cl) complying
APHA Max. 20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR